accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12080-32-9

IUPAC Name: platinum(2+) (1Z,5Z)-cycloocta-1,5-diene dichloride
Synonyms: platinum(2+) 1,5-cyclooctadiene, (Z,Z)- dichloride
Molecular Weight (g/mol): 374.17
Molecular Formula: C8H12Cl2Pt
InChi Key: VVAOPCKKNIUEEU-PHFPKPIQSA-L
SMILES: [Cl-].[Cl-].[Pt++].C1C\C=C/CC\C=C/1
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)