accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12081-16-2

IUPAC Name: tetrakis(ethene) dirhodium dichloride
Synonyms: chlorobis(ethylene)rhodium dimer
Molecular Weight (g/mol): 388.93
Molecular Formula: C8H16Cl2Rh2
InChi Key: QPOCZCJMFQWGSP-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Rh].[Rh].C=C.C=C.C=C.C=C
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)