CAS RN 121-44-8
Triethylamine, 99%, pure
CAS: 121-44-8 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, for HPLC, Fisher Chemical™
C6H15N, CAS Number-121-44-8, 250mL, 90 deg.C, CHEBI:35026, Colorless, 101.19g/mol, ZMANZCXQSJIPKH-UHFFFAOYSA-N, N,N-diethylethanamine, -115 deg.C, 9051, 101.193, Amber glass bottle, 12.4, Liquid, 8471, CCN(CC)CC, 3.5, 69mbar at 20 deg.C, 0.36 mPaS at 20 deg.C | CAS: 121-44-8 | C6H15N | 101.193 g/mol
Triethylamine, 99.7%, extra pure
CAS: 121-44-8 MDL Number: MFCD00009051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, 99%
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 MDL Number: MFCD00009051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, Extra Pure, SLR, Fisher Chemical™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 MDL Number: 9051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, Certified AR for Analysis, Fisher Chemical™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 MDL Number: 9051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, 99.5%, for analysis
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.19 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, 99+%
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 MDL Number: MFCD00009051 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, LC-MS Reagent, for general laboratory use, Solstice
Triethylamine, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N Synonym: triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Triethylamine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Triethylamine, for HPLC, ≥99.5%, Solstice
CAS: 121-44-8 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.19 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N IUPAC Name: triethylamine SMILES: CCN(CC)CC
- 1
- 2