accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 121-46-0

IUPAC Name: bicyclo[2.2.1]hepta-2,5-diene
Synonyms: norbornadiene
Molecular Weight (g/mol): 92.14
Molecular Formula: C7H8
InChi Key: SJYNFBVQFBRSIB-UHFFFAOYSA-N
SMILES: C1C2C=CC1C=C2
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results