CAS RN 121-47-1
CAS RN 121-47-1
IUPAC Name:
3-aminobenzene-1-sulfonic acid
Synonyms:
metanilic acid monosodium salt metanilic acid m-sulfanilic acid kyselina anilin-3-sulfonova aminobenzenesulfonic acid kyselina metanilova
Molecular Weight (g/mol):
173.19
Molecular Formula:
C6H7NO3S
InChi Key:
ZAJAQTYSTDTMCU-UHFFFAOYSA-N
SMILES:
NC1=CC=CC(=C1)S(O)(=O)=O
1
–
1
of
1
results
1
Metanilic acid, 98%
CAS: 121-47-1 Molecular Formula: C6H7NO3S Molecular Weight (g/mol): 173.19 MDL Number: MFCD00065345 InChI Key: ZAJAQTYSTDTMCU-UHFFFAOYSA-N Synonym: metanilic acid,m-sulfanilic acid,m-aminobenzenesulfonic acid,m-anilinesulfonic acid,1-aminobenzene-3-sulfonic acid,benzenesulfonic acid, 3-amino,aminobenzenesulfonic acid,kyselina metanilova,3-amino-benzenesulfonic acid,3-sulfoaniline PubChem CID: 8474 ChEBI: CHEBI:27764 IUPAC Name: 3-aminobenzenesulfonic acid SMILES: C1=CC(=CC(=C1)S(=O)(=O)O)N