accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SNNOO

CAS RN 121-66-4

IUPAC Name: 5-nitro-1,3-thiazol-2-amine
Synonyms: 2-thiazolamine, 5-nitro-
Molecular Weight (g/mol): 145.14
Molecular Formula: C3H3N3O2S
InChi Key: MIHADVKEHAFNPG-UHFFFAOYSA-N
SMILES: NC1=NC=C(S1)[N+]([O-])=O
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results