accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 121-73-3

IUPAC Name: 1-chloro-3-nitrobenzene
Synonyms: 3-nitrochlorobenzene
Molecular Weight (g/mol): 157.55
Molecular Formula: C6H4ClNO2
InChi Key: KMAQZIILEGKYQZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(Cl)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results