accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 121-86-8

IUPAC Name: 2-chloro-1-methyl-4-nitrobenzene
Synonyms: 2-chloro-4-nitrotoluene
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: LLYXJBROWQDVMI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results