accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONOHOOOH3CCH3CH3

CAS RN 1210863-93-6

IUPAC Name: 4-(acetyloxy)-1-[(tert-butoxy)carbonyl]pyrrolidine-3-carboxylic acid
Synonyms: 4-(acetyloxy)-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 273.29
Molecular Formula: C12H19NO6
InChi Key: OHQXZMNJROMWMW-UHFFFAOYNA-N
SMILES: CC(=O)OC1CN(CC1C(O)=O)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results