accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHN

CAS RN 1211-35-4

IUPAC Name: 2-(4-chlorophenyl)-1H-indole
Synonyms: 2-(4-chlorophenyl)-1H-indole
Molecular Weight (g/mol): 227.69
Molecular Formula: C14H10ClN
InChi Key: KDNXKQSAAZNUCK-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results