accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12112-67-3

IUPAC Name: cyclooctadiene iridium chloride dimer
Synonyms: cyclooctadiene iridium chloride dimer
Molecular Weight (g/mol): 671.7
Molecular Formula: C16H24Cl2Ir2
InChi Key: GGXFDBGPMSWGFD-MIXQCLKLSA-N
SMILES: [Cl].[Cl].[Ir].[Ir].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)