accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OI

CAS RN 121138-01-0

IUPAC Name: 3-iodooxolane
Synonyms: 3-iodooxolane
Molecular Weight (g/mol): 198
Molecular Formula: C4H7IO
InChi Key: BKIQORJIKOPRCG-UHFFFAOYNA-N
SMILES: IC1CCOC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)