accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12116-66-4

HCHCClClHf

CAS RN 12116-66-4

IUPAC Name: hafnium(4+) bis(cyclopenta-2,4-dien-1-ide) dichloride
Synonyms: hafnium(4+) bis(cyclopentadienide) dichloride
Molecular Weight (g/mol): 379.58
Molecular Formula: C10H10Cl2Hf
InChi Key: CSEGCHWAMVIXSA-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Hf+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)