accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 121195-03-7

CH3OONNHS

CAS RN 121195-03-7

IUPAC Name: ethyl 5-(thiophen-2-yl)-1H-pyrazole-3-carboxylate
Synonyms: ethyl 5-(thiophen-2-yl)-1H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 222.26
Molecular Formula: C10H10N2O2S
InChi Key: NTCRDZSJYGZRIE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NNC(=C1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results