CAS RN 1212-48-2
IUPAC Name:
hydrogen 2-benzyl-1H-1,3-benzodiazole chloride
Synonyms:
hydrogen bendazole chloride
Molecular Weight (g/mol):
244.72
Molecular Formula:
C14H13ClN2
InChi Key:
CJTQARUHALKPGG-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].C(C1=NC2=CC=CC=C2N1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Benzylbenzimidazole Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.