CAS RN 12120-15-9
IUPAC Name:
ethene bis(triphenylphosphane) platinum
Synonyms:
ethylene bis(triphenylphosphine) platinum
Molecular Weight (g/mol):
747.72
Molecular Formula:
C38H34P2Pt
InChi Key:
RZUASTIKPBCXPU-UHFFFAOYSA-N
SMILES:
[Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Ethylenebis(triphenylphosphine)platinum(0), 98%
CAS: 12120-15-9 Molecular Formula: C38H34P2Pt Molecular Weight (g/mol): 747.72 MDL Number: MFCD00012028 InChI Key: RZUASTIKPBCXPU-UHFFFAOYSA-N IUPAC Name: ethene bis(triphenylphosphane) platinum SMILES: [Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1