accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12125-01-8

IUPAC Name: ammonium hydrogen difluoride
Synonyms: ammonium fluoride
Molecular Weight (g/mol): 57.04
Molecular Formula: F2H5N
InChi Key: KVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILES: [H+].[NH4+].[F-].[F-]
Quantity 
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
Grade 
  • (4)
  • (7)
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (15)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (5)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (4)
  • (7)
  • (3)
  • (3)
  • (3)
  • (15)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)