accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 121325-67-5

IUPAC Name: imidazolidin-2-one
Synonyms: imidazolidinone monoethyleneurea 1,3-ethylene-urea N,N'-ethyleneurea ethyleneurea imidazolidinone (substance) ethylene urea N,N'-(1,2-ethanediyl)-urea 2-oxomidazolidine 1,3-ethyleneurea
Molecular Weight (g/mol): 86.09
Molecular Formula: C3H6N2O
InChi Key: YAMHXTCMCPHKLN-UHFFFAOYSA-N
SMILES: O=C1NCCN1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)