accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1214-39-7

IUPAC Name: N-benzyl-7H-purin-6-amine
Synonyms: 6-benzylaminopurine
Molecular Weight (g/mol): 225.26
Molecular Formula: C12H11N5
InChi Key: NWBJYWHLCVSVIJ-UHFFFAOYSA-N
SMILES: C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (9)
Quantity 
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (9)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)