accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

IrIrOOCH3CH3

CAS RN 12148-71-9

IUPAC Name: bis(cycloocta-1,5-diene) bis(methyloxidanium) diiridium
Synonyms: bis(1,5-cyclooctadiene) bis(methyloxidanium) diiridium
Molecular Weight (g/mol): 666.9
Molecular Formula: C18H34Ir2O2
InChi Key: MNKCGUKVRJZKEQ-UHFFFAOYSA-P
SMILES: [Ir].[Ir].C[OH2+].C[OH2+].C1CC=CCCC=C1.C1CC=CCCC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results