CAS RN 1215681-42-7
CAS RN 1215681-42-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Imipramine N-Oxide Hydrate, TRC
CAS: 1215681-42-7 Molecular Formula: C19H24N2O • x(H2O) Molecular Weight (g/mol): 296.411802 Synonym: 10,11-Dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine N-Oxide Hydrate,5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine 5-oxide Hydrate,Imipraminoxide Hydrate IUPAC Name: 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine oxide;hydrate SMILES: CN(CCCN1C2=C(C=CC=C2)CCC3=C1C=CC=C3)(C)=O