CAS RN 1217036-71-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,6-Dihydroxypyrazolo[3,4-d]pyrimidine-13C,15N2, TRC
CAS: 1217036-71-9 Molecular Formula: 13C C4 H4 15N2 N2 O2 Molecular Weight (g/mol): 155.09 Synonym: Oxypurinol-13C,15N2,Oxypurinol-3-13C-1,2-15N2,Oxipurinol-13C,15N2,1H-Pyrazolo[3,4-d]pyrimidine-4,6(5H,7H)-dione-3-13C-1,2-15N2,1H-Pyrazolo[3,4-d]pyrimidin-4,6-diol-3-13C-1,2-15N2,4,6-Dihydroxypyrazolo[3,4-d]pyrimidine-3-13C-1,2-15N2,4,6-Dioxopyrazolo[3,4-d]pyrimidine-3-13C-1,2-15N2,4,6-Dihydroxypyrazolo[3,4-d]pyrimidine-13C,15N2 IUPAC Name: 1,2-dihydro(5^{13}C,1,2-^{15}N_{2})pyrazolo[3,4-d]pyrimidine-4,6-dione SMILES: O=C1NC(=O)C2=[13CH][15NH][15NH]C2=N1