accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 121808-62-6

OHO(R)SNO(S)OHN

CAS RN 121808-62-6

IUPAC Name: (4R)-3-[(2S)-5-oxopyrrolidine-2-carbonyl]-1,3-thiazolidine-4-carboxylic acid
Synonyms: pidotimod
Molecular Weight (g/mol): 244.27
Molecular Formula: C9H12N2O4S
InChi Key: UUTKICFRNVKFRG-WDSKDSINSA-N
SMILES: OC(=O)[C@@H]1CSCN1C(=O)[C@@H]1CCC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)