accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHNH2OFFF

CAS RN 1218228-20-6

IUPAC Name: 2-amino-3-[2-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 2-amino-3-[2-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 249.19
Molecular Formula: C10H10F3NO3
InChi Key: ILPKLBNKYRAICX-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC=C1OC(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)