accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1218790-44-3

IUPAC Name: 1-{[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}piperidine
Synonyms: 1-{[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}piperidine
Molecular Weight (g/mol): 307.26
Molecular Formula: C16H26BNO2S
InChi Key: KDPVLCCGXRCQCV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCCC2)S1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results