accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3CNOOCH3(E/Z)

CAS RN 1218790-51-2

IUPAC Name: ethyl 2-cyano-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoate
Synonyms: ethyl 2-cyano-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoate
Molecular Weight (g/mol): 327.19
Molecular Formula: C18H22BNO4
InChi Key: DNALFJBPPFISJQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results