accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1218791-38-8

IUPAC Name: 1-{4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazin-1-yl}ethan-1-one
Synonyms: 1-{4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazin-1-yl}ethanone
Molecular Weight (g/mol): 330.24
Molecular Formula: C18H27BN2O3
InChi Key: VRZVSHHLWMAHAZ-UHFFFAOYSA-N
SMILES: CC(=O)N1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results