accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12196-72-4

IUPAC Name: lanthanum pentanickel
Synonyms: lanthanum pentanickel
Molecular Weight (g/mol): 432.37
Molecular Formula: LaNi5
InChi Key: BUCVQDCWULHLJZ-UHFFFAOYSA-N
SMILES: [Ni].[Ni].[Ni].[Ni].[Ni].[La]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Form 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results