accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3O

CAS RN 122-03-2

IUPAC Name: 4-(propan-2-yl)benzaldehyde
Synonyms: cuminaldehyde cumic aldehyde cuminyl aldehyde P-isopropyl benzaldehyde cuminal cumaldehyde P-cuminic aldehyde cuminic aldehyde
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: WTWBUQJHJGUZCY-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=O)C=C1
Boiling Point 
  • (7)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (7)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)