accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 122-20-3

IUPAC Name: 1-[bis(2-hydroxypropyl)amino]propan-2-ol
Synonyms: triisopropanolamine
Molecular Weight (g/mol): 191.27
Molecular Formula: C9H21NO3
InChi Key: SLINHMUFWFWBMU-UHFFFAOYNA-N
SMILES: CC(O)CN(CC(C)O)CC(C)O
Boiling Point 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)