accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNH

CAS RN 122-37-2

IUPAC Name: 4-(phenylamino)phenol
Synonyms: phenol, 4-(phenylamino)-
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: JTTMYKSFKOOQLP-UHFFFAOYSA-N
SMILES: OC1=CC=C(NC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)