CAS RN 122-40-7
IUPAC Name:
(2Z)-2-(phenylmethylidene)heptanal
Synonyms:
amyl cinnamic aldehyde amylcinnamaldehyde pentylcinnamaldehyde jasmine aldehyde α-amylcinnamaldehyde (natural) flomine amylcinnamic acid aldehyde jasminal jasminaldehyde α-amylcinnamicaldehyde
Molecular Weight (g/mol):
202.3
Molecular Formula:
C14H18O
InChi Key:
HMKKIXGYKWDQSV-KAMYIIQDSA-N
SMILES:
CCCCC\C(C=O)=C\C1=CC=CC=C1
1
–
2
of
2
results
1
Alpha-Amylcinnamaldehyde stabilized with BHT, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
a-Amylcinnamaldehyde stabilized with BHT, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.