accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 122-78-1

IUPAC Name: 2-phenylacetaldehyde
Synonyms: phenylacetaldehyde phenylacetic aldehyde α-tolualdehyde α-toluic aldehyde phenacetaldehyde phenylacetaldehyde (natural)
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: DTUQWGWMVIHBKE-UHFFFAOYSA-N
SMILES: O=CCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results