accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 122-80-5

IUPAC Name: N-(4-aminophenyl)acetamide
Synonyms: P-aminoacetanilide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: CHMBIJAOCISYEW-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)