accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 122-85-0

IUPAC Name: N-(4-formylphenyl)acetamide
Synonyms: P-formylacetanilide
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: SKLUWKYNZNXSLX-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results