accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNNOH

CAS RN 122-96-3

IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethan-1-ol
Synonyms: 1,4-piperazinediethanol
Molecular Weight (g/mol): 174.24
Molecular Formula: C8H18N2O2
InChi Key: VARKIGWTYBUWNT-UHFFFAOYSA-N
SMILES: OCCN1CCN(CCO)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results