accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 122-99-6

IUPAC Name: 2-phenoxyethan-1-ol
Synonyms: phenoxyethanol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: QCDWFXQBSFUVSP-UHFFFAOYSA-N
SMILES: OCCOC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (4)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (5)
Boiling Point 
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (5)
  • (3)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)