CAS RN 122008-85-9
IUPAC Name:
butyl (2R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propanoate
Synonyms:
butyl (2R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propanoate
Molecular Weight (g/mol):
357.38
Molecular Formula:
C20H20FNO4
InChi Key:
TYIYMOAHACZAMQ-CQSZACIVSA-N
SMILES:
CCCCOC(=O)[C@@H](C)OC1=CC=C(OC2=CC=C(C=C2F)C#N)C=C1
1
–
3
of
3
results
1
Cyhalofop-butyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Cyhalofop-butyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Cyhalofop-butyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.