accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNH3COHO

CAS RN 122222-09-7

IUPAC Name: 1,2-dimethyl-1H-imidazole-5-carboxylic acid
Synonyms: 2,3-dimethylimidazole-4-carboxylic acid
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: GHSZJLADCHKTGG-UHFFFAOYSA-N
SMILES: CN1C(C)=NC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results