accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1222709-28-5

NHOONH2HCl

CAS RN 1222709-28-5

IUPAC Name: benzyl N-(3-aminocyclohexyl)carbamate hydrochloride
Synonyms: benzyl N-(3-aminocyclohexyl)carbamate hydrochloride
Molecular Weight (g/mol): 284.78
Molecular Formula: C14H21ClN2O2
InChi Key: SBODKRLYZILWEM-UHFFFAOYNA-N
SMILES: Cl.NC1CCCC(C1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results