accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12233-34-0

OTiOTiOOTiOOBiOOTiOOBiO

CAS RN 12233-34-0

IUPAC Name: tetraoxo-2,4,6,8,10-pentaoxa-1,11-dibisma-3,5,7,9-tetratitanaundecane-1,11-dione
Synonyms: tetraoxo-2,4,6,8,10-pentaoxa-1,11-dibisma-3,5,7,9-tetratitanaundecane-1,11-dione
Molecular Weight (g/mol): 785.42
Molecular Formula: Bi2O11Ti4
InChi Key: TZQKEPWGUIJJIY-UHFFFAOYSA-N
SMILES: O=[Bi]O[Ti](=O)O[Ti](=O)O[Ti](=O)O[Ti](=O)O[Bi]=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results