accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3OONO

CAS RN 1223573-41-8

IUPAC Name: tert-butyl 1-oxa-6-azaspiro[3.3]heptane-6-carboxylate
Synonyms: tert-butyl 1-oxa-6-azaspiro[3.3]heptane-6-carboxylate
Molecular Weight (g/mol): 199.25
Molecular Formula: C10H17NO3
InChi Key: QOXFRPSIASGKGE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC2(CCO2)C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results