accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12244-57-4

IUPAC Name: gold(1+) disodium 2-sulfanidylbutanedioate hydrate
Synonyms: sodium aurothiomalate
Molecular Weight (g/mol): 408.09
Molecular Formula: C4H5AuNa2O5S
InChi Key: YLQOAPBVYJCTPW-UHFFFAOYNA-K
SMILES: O.[Na+].[Na+].[Au+].[O-]C(=O)CC([S-])C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results