accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 12260-45-6

OHCs

CAS RN 12260-45-6

IUPAC Name: caesium(1+) hydrate hydroxide
Synonyms: caesium(1+) hydrate hydroxide
Molecular Weight (g/mol): 167.93
Molecular Formula: CsH3O2
InChi Key: ABSOMGPQFXJESQ-UHFFFAOYSA-M
SMILES: O.[OH-].[Cs+]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results