accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3OHNHSCH3OOH3CNCH3OHNHSCH3OOOHOOHO(E)

CAS RN 122647-32-9

IUPAC Name: (2E)-but-2-enedioic acid; bis(N-(4-{4-[ethyl(heptyl)amino]-1-hydroxybutyl}phenyl)methanesulfonamide)
Synonyms: ibutilide
Molecular Weight (g/mol): 885.23
Molecular Formula: C44H76N4O10S2
InChi Key: PCIOHQNIRPWFMV-WXXKFALUNA-N
SMILES: OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)