accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 122666-87-9

(S)ONH2OONaNaH2OH2O

CAS RN 122666-87-9

IUPAC Name: 2-amino-3-(4-hydroxyphenyl)propanoic acid dihydrate disodium
Synonyms: tyrosin dihydrate disodium
Molecular Weight (g/mol): 263.2
Molecular Formula: C9H15NNa2O5
InChi Key: FBKIASNRVHFWNA-UHFFFAOYNA-N
SMILES: O.O.[Na].[Na].NC(CC1=CC=C(O)C=C1)C(O)=O
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)