accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCH3NH3CCH3SNCl

CAS RN 122965-43-9

IUPAC Name: 7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium hydrate chloride
Synonyms: 7-(dimethylamino)-N,N-dimethylphenothiazin-3-iminium hydrate chloride
Molecular Weight (g/mol): 337.87
Molecular Formula: C16H20ClN3OS
InChi Key: WQVSELLRAGBDLX-UHFFFAOYSA-M
SMILES: O.[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Color 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Grade 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (5)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)