accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C

CAS RN 123-01-3

IUPAC Name: dodecylbenzene
Synonyms: dodecylbenzene marlican phenyldodecan dodecylbenzene, crude crude dodecylbenzene ucane alkylate 12 dodecylbenzene (linear) phenyldodecane nalkylene 500 alkylate P 1
Molecular Weight (g/mol): 246.44
Molecular Formula: C18H30
InChi Key: KWKXNDCHNDYVRT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (6)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)