accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 123-29-5

IUPAC Name: ethyl nonanoate
Synonyms: ethyl nonanoate
Molecular Weight (g/mol): 186.3
Molecular Formula: C11H22O2
InChi Key: BYEVBITUADOIGY-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)OCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results