accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOH

CAS RN 123-31-9

IUPAC Name: benzene-1,4-diol
Synonyms: α-hydroquinone hydroquinone product pyrogentistic acid melpaque hydroquinone, monocopper (2+) salt hydroquinone gr phiaquin hidroquilaude vinas brand of hydroquinone robins brand of hydroquinone
Molecular Weight (g/mol): 110.11
Molecular Formula: C6H6O2
InChi Key: QIGBRXMKCJKVMJ-UHFFFAOYSA-N
SMILES: OC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (11)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (8)
Percent Purity 
  • (1)
  • (6)
  • (5)
Physical Form 
  • (8)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (11)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (8)
  • (1)
  • (6)
  • (5)
  • (8)
  • (3)
  • (1)